4-Fluor-3-methylbenzylalkohol

Lieferant: FLUOROCHEM

Synonyme: (4-Fluoro-3-methylphenyl)methanol

005794-1G 005794-5G 005794-10G
FLUO005794-1GEA 69.9 EUR
FLUO005794-1G FLUO005794-5G FLUO005794-10G
4-Fluor-3-methylbenzylalkohol
4-Fluor-3-methylbenzylalkohol
Formel: C₈H₉FO
Molecular Weight: 140.16 g/mol
MDL: MFCD01631427
CAS-Nummer: 261951-66-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR