5-Acetyl-2,4-dimethylthiazol ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,4-Dimethyl-5-acetylthiazol, 1-(2,4-Dimethylthiazol-5-yl)ethan-1-on
A13741.06
A13741.14
A13741.06EA
21.4
EUR
A13741.06
A13741.14
5-Acetyl-2,4-dimethylthiazol ≥99%
5-Acetyl-2,4-dimethylthiazol
Purity: 99%
CAS No.: 38205-60-6
Molecular Formula: C7H9NOS
Formula Weight: 155.22
Flash Point: 104°(219°F)
Refractive Index: 1.5430
Boiling Point: 230°
Beilstein Reference: 120560
MDL No.: MFCD00005331
Formel:
C₇H₉NOS Molecular Weight: 155,22 g/mol Siedepunkt: 230 °C Dichte: 1,150 Flammpunkt: 104 °C (219 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005331 CAS-Nummer: 38205-60-6 EINECS: 253-826-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...