5-Acetyl-2,4-dimethylthiazol

Lieferant: FLUOROCHEM

Synonyme: 2,4-Dimethyl-5-acetylthiazol, 1-(2,4-Dimethylthiazol-5-yl)ethan-1-on

002762-5G 002762-25G
FLUO002762-5GEA 0 EUR
FLUO002762-5G FLUO002762-25G
5-Acetyl-2,4-dimethylthiazol
5-Acetyl-2,4-dimethylthiazol
Formel: C₇H₉NOS
Molecular Weight: 155.22 g/mol
Siedepunkt: 228
Dichte: 1.15 g/cm³ (20 °C)
MDL: MFCD00005331
CAS-Nummer: 38205-60-6
EINECS: 253-826-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR