2-Chlor-4-fluorphenylboronsäure ≥98%

Lieferant: Thermo Scientific
Achtung

H27591.14 H27591.06 H27591.03
H27591.14EA 669 EUR
H27591.14 H27591.06 H27591.03
2-Chlor-4-fluorphenylboronsäure ≥98%
2-Chlor-4-fluorphenylboronsäure

Purity: 98% CAS No.: 313545-72-1 Molecular Formula: C6H5BClFO2 Formula Weight: 174.37 MDL No.: MFCD02684295

Formel: C₆H₅BClFO₂
Molecular Weight: 174,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684295
CAS-Nummer: 313545-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR