2-Chlor-5-(trifluormethyl)phenylboronsäure 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2-Chlor-5-(trifluormethyl)phenyl)metaborsäure, Dihydroxy(2-chlor-5-(trifluormethyl)phenyl)boran, 2-Chlor-5-(trifluormethyl)benzolboronsäure

L20104.03 L20104.06
L20104.03EA 46.8 EUR
L20104.03 L20104.06
2-Chlor-5-(trifluormethyl)phenylboronsäure 96%
2-Chlor-5-(trifluormethyl)phenylboronsäure

Purity: 96% CAS No.: 182344-18-9 Molecular Formula: C7H5BClF3O2 Formula Weight: 224.38 MDL No.: MFCD00797335

Formel: C₇H₅BClF₃O₂
Molecular Weight: 224.37
Lagertemperatur: Raumtemperatur
MDL: MFCD00797335
CAS-Nummer: 182344-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR