2-Acetylphenylboronsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Acetylbenzolboronsäure, Dihydroxy(2-acetylphenyl)boran, (2-Acetylphenyl)metaborsäure
H27327.14
H27327.06
H27327.14EA
766
EUR
H27327.14
H27327.06
2-Acetylphenylboronsäure ≥97%
2-Acetylphenylboronsäure
Purity: 97%
CAS No.: 308103-40-4
Molecular Formula: C8H9BO3
Formula Weight: 163.97
Melting Point: ca 170° dec.
MDL No.: MFCD01321263
Formel:
C₈H₉BO₃ Molecular Weight: 163,97 g/mol Schmelzpunkt: ∼ 170 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01321263 CAS-Nummer: 308103-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...