Eisen(III)citrat, Sigma-Aldrich®

Lieferant: Merck

F6129-1KG F6129-250G
SIALF6129-1KGEA 289 EUR
SIALF6129-1KG SIALF6129-250G
Eisen(III)citrat, Sigma-Aldrich®
Eisen(III)citrat
Formel: C₆H₅FeO₇
Molecular Weight: 244.95 g/mol
CAS-Nummer: 28633-45-6
EINECS: 249-117-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR