2-Tolylboronsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Methylbenzolboronsäure, 2-Methylphenylboronsäure, (2-Methylphenyl)metaborsäure, Dihydroxy(2-methylphenyl)boran

B23154.03 B23154.06
B23154.03EA 39.6 EUR
B23154.03 B23154.06
2-Tolylboronsäure ≥98%
2-Tolylboronsäure

Purity: 98% CAS No.: 16419-60-6 Molecular Formula: C7H9BO2 Formula Weight: 135.96 Melting Point: ca 162-164° Beilstein Reference: 2935796

Formel: C₇H₉BO₂
Molecular Weight: 135,96 g/mol
Schmelzpunkt: ∼ 162…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00093526
CAS-Nummer: 16419-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR