2-Tolylboronsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Methylbenzolboronsäure, 2-Methylphenylboronsäure, (2-Methylphenyl)metaborsäure, Dihydroxy(2-methylphenyl)boran

309260010 AC30926.10 309260050
ACRO309260010EA 30.6 EUR
ACRO309260010 MAYBAC30926.10 ACRO309260050
2-Tolylboronsäure 95%
2-Tolylboronsäure
Formel: C₇H₉BO₂
Molecular Weight: 135,96 g/mol
Schmelzpunkt: 164…165 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00093526
CAS-Nummer: 16419-60-6

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder or crystals
Infrared spectrum Conforms
Melting point 160 to 166 °C
Additional info May contain varying amounts of anhydrides
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR