2-Tolylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Methylbenzolboronsäure, 2-Methylphenylboronsäure, (2-Methylphenyl)metaborsäure, Dihydroxy(2-methylphenyl)boran

002464-5G 002464-25G 002464-1G
FLUO002464-5GEA 36.9 EUR
FLUO002464-5G FLUO002464-25G FLUO002464-1G
2-Tolylboronsäure
2-Tolylboronsäure
Formel: C₇H₉BO₂
Molecular Weight: 135.96 g/mol
Schmelzpunkt: 160…166 °C
MDL: MFCD00093526
CAS-Nummer: 16419-60-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR