2-Noninsäuremethylester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl-2-noninoat

M-Clarity™ Qualitätsstufe = MQ300

W272604-100G-K W272604-1KG-K
SIALW272604-100G-KEA 147 EUR
SIALW272604-100G-K SIALW272604-1KG-K
2-Noninsäuremethylester, Sigma-Aldrich®
2-Nonynoic acid methyl ester
Formel: C₁₀H₁₆O₂
Molecular Weight: 168.24 g/mol
MDL: MFCD00009547
CAS-Nummer: 111-80-8
EINECS: 203-909-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR