2-Phenoxynicotinsäure
Lieferant: Thermo Scientific
BTB02988.1
BTB02988.10
BTB02988.50
MAYBBTB02988.1EA
0
EUR
MAYBBTB02988.1
MAYBBTB02988.10
MAYBBTB02988.50
2-Phenoxynicotinsäure
2-Phenoxynicotinsäure
Formel:
C₁₂H₉NO₃ |
MDL:
MFCD00014629 CAS-Nummer: 35620-71-4 EINECS: 252-647-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...