3-Amino-4-ethylcapronsäure
Lieferant: BIONET RESEARCH
8T-0051.500
8T-0051.1
8T-0051.5
8T-0051.10
BIOT8T-0051.500EA
0
EUR
BIOT8T-0051.500
BIOT8T-0051.1
BIOT8T-0051.5
BIOT8T-0051.10
3-Amino-4-ethylcapronsäure
3-Amino-4-ethylcapronsäure
Formel:
C₈H₁₇NO₂ |
MDL:
MFCD01863280 CAS-Nummer: 204191-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...