3-Amino-5-methylcapronsäure

Lieferant: BIONET RESEARCH

Synonyme: 3-Amino-5-methylhexansäure

8T-0019.500 8T-0019.1 8T-0019.5 8T-0019.10
BIOT8T-0019.500EA 0 EUR
BIOT8T-0019.500 BIOT8T-0019.1 BIOT8T-0019.5 BIOT8T-0019.10
3-Amino-5-methylcapronsäure
3-Amino-5-methylcapronsäure
Formel: C₇H₁₅NO₂
Molecular Weight: 145,2 g/mol
Schmelzpunkt: 227 °C
MDL: MFCD01863281
CAS-Nummer: 3653-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR