3-Aminophenylboronsäure Hemisulfat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (3-Aminophenyl)boronic acid sulphate (2:1), 3-Aminobenzeneboronic acid hemisulphate

A17240.03 A17240.06 A17240.14
A17240.03EA 44.6 EUR
A17240.03 A17240.06 A17240.14
3-Aminophenylboronsäure Hemisulfat ≥98%
3-Aminophenylboronsäure Hemisulfat

Purity: 98+% CAS No.: 66472-86-4 Molecular Formula: C6H8BNO2.0.5H2SO4 Formula Weight: 185.98 Melting Point: 300° Beilstein Reference: 8094151

Formel: C₁₂H₁₈B₂N₂O₈S
Molecular Weight: 371.97 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013111
CAS-Nummer: 66472-86-4
EINECS: 266-376-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR