3-Aminophenylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
Dihydroxy(3-aminophenyl)boran, 3-Aminobenzolboronsäure, (3-Aminophenyl)metaborsäure
021277-5G
021277-25G
FLUO021277-5GEA
76.2
EUR
FLUO021277-5G
FLUO021277-25G
3-Aminophenylboronsäure
3-Aminophenylboronsäure
Formel:
C₆H₈BNO₂ Molecular Weight: 136.95 g/mol |
MDL:
MFCD00007755 CAS-Nummer: 30418-59-8 EINECS: 250-189-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...