3-Aminophenylboronsäure

Lieferant: CHEMPUR

Synonyme: Dihydroxy(3-aminophenyl)boran, 3-Aminobenzolboronsäure, (3-Aminophenyl)metaborsäure

A02-M130.2 A02-M130.10
CHMPA02-M130.2EA 0 EUR
CHMPA02-M130.2 CHMPA02-M130.10
3-Aminophenylboronsäure
3-Aminophenylboronsäure
Formel: C₆H₈BNO₂
Molecular Weight: 136.95 g/mol
MDL: MFCD00007755
CAS-Nummer: 30418-59-8
EINECS: 250-189-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR