3-Chlor-4-fluorphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Dihydroxy(3-chlor-4-fluorphenyl)boran, (3-Chlor-4-fluorphenyl)metaborsäure, 3-Chlor-4-fluorbenzolboronsäure
B22755.06
B22755.14
B22755.06EA
80.2
EUR
B22755.06
B22755.14
3-Chlor-4-fluorphenylboronsäure ≥98%
3-Chlor-4-fluorphenylboronsäure
Purity: 98%
CAS No.: 144432-85-9
Molecular Formula: C6H5BClFO2
Formula Weight: 174.37
Melting Point: 255-260°
Beilstein Reference: 7369789
Formel:
C₆H₅BClFO₂ Molecular Weight: 174,37 g/mol Schmelzpunkt: 255…260 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051800 CAS-Nummer: 144432-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...