3-Acetylphenylboronsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(3-Acetylphenyl)metaborsäure, Dihydroxy(3-acetylphenyl)boran, 3-Acetylbenzolboronsäure
B23478.03
B23478.06
B23478.03EA
24.5
EUR
B23478.03
B23478.06
3-Acetylphenylboronsäure ≥97%
3-Acetylphenylboronsäure
Purity: 97%
CAS No.: 204841-19-0
Molecular Formula: C8H9BO3
Formula Weight: 163.97
Melting Point: ca 200-205°
Beilstein Reference: 8255673
Formel:
C₈H₉BO₃ Molecular Weight: 163,97 g/mol Schmelzpunkt: ∼ 200…205 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01074678 CAS-Nummer: 204841-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...