3-Iodphenylboronsäure

Lieferant: FLUOROCHEM

011041-5G 011041-1G
FLUO011041-5GEA 201 EUR
FLUO011041-5G FLUO011041-1G
3-Iodphenylboronsäure
3-Iodphenylboronsäure
Formel: C₆H₆BIO₂
MDL: MFCD01319015
CAS-Nummer: 221037-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR