3-Tolylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Methylbenzolboronsäure, Dihydroxy(3-methylphenyl)boran, (3-Methylphenyl)metaborsäure, 3-Methylphenylboronsäure

B23025.06 B23025.03 B23025.14
B23025.06EA 96.1 EUR
B23025.06 B23025.03 B23025.14
3-Tolylboronsäure ≥97%
3-Tolylboronsäure

Purity: 97% CAS No.: 17933-03-8 Molecular Formula: C7H9BO2 Formula Weight: 135.96 Melting Point: ca 160-162° Beilstein Reference: 2935968

Formel: C₇H₉BO₂
Molecular Weight: 135,96 g/mol
Schmelzpunkt: ∼ 160…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040198
CAS-Nummer: 17933-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR