3-Tolylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
3-Methylbenzolboronsäure, Dihydroxy(3-methylphenyl)boran, (3-Methylphenyl)metaborsäure, 3-Methylphenylboronsäure
010853-5G
010853-25G
FLUO010853-5GEA
0
EUR
FLUO010853-5G
FLUO010853-25G
3-Tolylboronsäure
3-Tolylboronsäure
Formel:
C₇H₉BO₂ Molecular Weight: 135,96 g/mol Schmelzpunkt: 165 °C |
MDL:
MFCD00040198 CAS-Nummer: 17933-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...