Cytosin 99+%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-2-pyrimidinol, 4-Amino-2-hydroxypyrimidin

161760050 161760250 161761000
ACRO161760050EA 124 EUR
ACRO161760050 ACRO161760250 ACRO161761000
Cytosin 99+%
Cytosin
Formel: C₄H₅N₃O
Molecular Weight: 111,1 g/mol
Schmelzpunkt: >300 °C
Dichte: 0,99771 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006034
CAS-Nummer: 71-30-7
EINECS: 200-749-5
Merck Index: 13,02824

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 5 hrs)
Sulfated ash ≤0.1 % (800°C)
HPLC ≥99.0 %
UV spectrophotometry molar absorptivity 10400 +/- 2%, on dry basis
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR