4-Aminonicotinsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Aminopyridin-3-carbonsäure, 4-Amino-3-pyridincarbonsäure
361130010
361130050
ACRO361130010EA
27.7
EUR
ACRO361130010
ACRO361130050
4-Aminonicotinsäure 97%
4-Aminonicotinsäure
Formel:
C₆H₆N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234183 CAS-Nummer: 7418-65-7 |
Spezifikation Testergebnisse
Appearance | White to yellow to beige Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...