4-Chlor-2-(trifluormethyl)phenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Chloro-2-(trifluoromethyl)benzeneboronic acid, (4-Chloro-2-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-chloro-2-(trifluoromethyl)phenyl)borane

H28351.09 H28351.03
H28351.09EA 226 EUR
H28351.09 H28351.03
4-Chlor-2-(trifluormethyl)phenylboronsäure ≥97%
4-Chlor-2-(trifluormethyl)phenylboronsäure

Purity: 97% CAS No.: 313545-41-4 Molecular Formula: C7H5BClF3O2 Formula Weight: 224.38 Melting Point: 180-182° MDL No.: MFCD04973086

Formel: C₇H₅BClF₃O₂
Molecular Weight: 224.37
Schmelzpunkt: 180…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04973086
CAS-Nummer: 313545-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR