4-Chlornicotinsäure

Lieferant: CHEMPUR

Synonyme: 4-Chlor-3-pyridincarbonsäure, 4-Chlorpyridin-3-carbonsäure

FL13136.POR 013136-5G 013136-1G
CHMPFL13136.POREA 0 EUR
CHMPFL13136.POR FLUO013136-5G FLUO013136-1G
4-Chlornicotinsäure
4-Chlornicotinsäure
Formel: C₆H₄ClNO₂
Molecular Weight: 157,56 g/mol
MDL: MFCD00128860
CAS-Nummer: 10177-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR