4-Chlornicotinsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Chlor-3-pyridincarbonsäure, 4-Chlorpyridin-3-carbonsäure
L20029.03
L20029.06
L20029.03EA
81.4
EUR
L20029.03
L20029.06
4-Chlornicotinsäure ≥97%
4-Chlornicotinsäure
Purity: 97%
CAS No.: 10177-29-4
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Melting Point: ca 165° dec.
Beilstein Reference: 9255849
Formel:
C₆H₄ClNO₂ Molecular Weight: 157,56 g/mol Schmelzpunkt: ∼ 165 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128860 CAS-Nummer: 10177-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...