4-Acetylphenylboronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane

369400010 369400050 369400250
ACRO369400010EA 26.7 EUR
ACRO369400010 ACRO369400050 ACRO369400250
4-Acetylphenylboronsäure 97%
4-Acetylphenylboronsäure
Formel: C₈H₉BO₃
Molecular Weight: 163,97 g/mol
Schmelzpunkt: 243…247 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01074667
CAS-Nummer: 149104-90-5

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR