4-Acetylphenylboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane
M-Clarity™ Qualitätsstufe = MQ100
470821-25G
470821-5G
SIAL470821-25GEA
284
EUR
SIAL470821-25G
SIAL470821-5G
4-Acetylphenylboronsäure, Sigma-Aldrich®
4-Acetylphenylboronsäure
Formel:
C₈H₉BO₃ Molecular Weight: 163,97 g/mol |
MDL:
MFCD01074667 CAS-Nummer: 149104-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...