4-Vinylphenylboronsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Dihydroxy(4-vinylphenyl)boran, (4-Vinylphenyl)metaborsäure, 4-Vinylbenzolboronsäure
B23709.03
B23709.06
B23709.14
B23709.03EA
36
EUR
B23709.03
B23709.06
B23709.14
4-Vinylphenylboronsäure ≥97%
4-Vinylphenylboronsäure
Purity: 97%
CAS No.: 2156-04-9
Molecular Formula: C8H9BO2
Formula Weight: 147.97
Storage Temperature: 2°C to 8°C
Melting Point: ca 190°
Beilstein Reference: 2829387
MDL No.: MFCD00239441
Formel:
C₈H₉BO₂ Molecular Weight: 147,97 g/mol Schmelzpunkt: ∼ 190 °C |
MDL:
MFCD00239441 CAS-Nummer: 2156-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...