4-Vinylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dihydroxy(4-vinylphenyl)boran, (4-Vinylphenyl)metaborsäure, 4-Vinylbenzolboronsäure

417580-1G 417580-5G
SIAL417580-1GEA 59.6 EUR
SIAL417580-1G SIAL417580-5G
4-Vinylphenylboronsäure, Sigma-Aldrich®
4-Vinylphenylboronsäure
Formel: C₈H₉BO₂
Molecular Weight: 147,97 g/mol
MDL: MFCD00239441
CAS-Nummer: 2156-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR