4-Iodphenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Jodbenzolboronsäure, 4-Iodbenzolboronsäure, (4-Iodphenyl)metaborsäure, (4-Jodphenyl)metaborsäure, Dihydroxy(4-jodphenyl)boran, Dihydroxy(4-iodphenyl)boran
H51119.03
H51119.06
H51119.14
H51119.03EA
30.9
EUR
H51119.03
H51119.06
H51119.14
4-Iodphenylboronsäure ≥97%
4-Iodphenylboronsäure
Purity: 97%
CAS No.: 5122-99-6
Molecular Formula: C6H6BIO2
Formula Weight: 247.83
MDL No.: MFCD02093073
Formel:
C₆H₆BIO₂ Molecular Weight: 247.83 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319014 CAS-Nummer: 5122-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...