4-Tolylboronsäure

Lieferant: FLUOROCHEM

010854-100G 010854-25G 010854-5G
FLUO010854-100GEA 143 EUR
FLUO010854-100G FLUO010854-25G FLUO010854-5G
4-Tolylboronsäure
4-Tolylboronsäure
Formel: C₇H₉BO₂
Schmelzpunkt: 260 °C
MDL: MFCD00039138
CAS-Nummer: 5720-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR