5,6-Dichlornicotinsäure

Lieferant: BIONET RESEARCH

Synonyme: 5,6-Dichlorpyridin-3-carbonsäure, 5,6-Dichlor-3-pyridincarbonsäure

12X-0700.1 12X-0700.10 12X-0700.5 12X-0700.500
BIOT12X-0700.1EA 0 EUR
BIOT12X-0700.1 BIOT12X-0700.10 BIOT12X-0700.5 BIOT12X-0700.500
5,6-Dichlornicotinsäure
5,6-Dichlornicotinsäure
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192 g/mol
MDL: MFCD00075181
CAS-Nummer: 41667-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR