5,6-Dichlornicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 5,6-Dichlorpyridin-3-carbonsäure, 5,6-Dichlor-3-pyridincarbonsäure

010699-5G 010699-25G
FLUO010699-5GEA 34.1 EUR
FLUO010699-5G FLUO010699-25G
5,6-Dichlornicotinsäure
5,6-Dichlornicotinsäure
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192 g/mol
MDL: MFCD00075181
CAS-Nummer: 41667-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR