δ-Valerolacton ≥98% may contain polymer
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Pentansäure-δ-lacton, 5-Hydroxyvaleriansäurelacton, Tetrahydro-2H-pyran-2-on, Pentan-5-olid
L04794.14
L04794.22
L04794.36
L04794.14EA
54.3
EUR
L04794.14
L04794.22
L04794.36
δ-Valerolacton ≥98% may contain polymer
δ-Valerolacton
Purity: 98%
CAS No.: 542-28-9
Molecular Formula: C5H8O2
Formula Weight: 100.12
Storage Temperature: 2°C to 8°C
Flash Point: 112°(233°F)
Refractive Index: 1.4580
Melting Point: -13 to -12°
Boiling Point: 58-60°/0.5mm
Beilstein Reference: 106436
MDL No.: MFCD00006645
Formel:
C₅H₈O₂ Molecular Weight: 100.12 g/mol Siedepunkt: 58…60 °C (0.5 mmHg) Schmelzpunkt: –13 °C…–12 °C Dichte: 1.056 Flammpunkt: 112 °C (233 °F) |
MDL:
MFCD00006645 CAS-Nummer: 542-28-9 EINECS: 208-807-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...