5-Methylnicotinsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Methyl-3-pyridincarbonsäure, 5-Methylpyridin-3-carbonsäure
L16370.03
L16370.06
L16370.03EA
104
EUR
L16370.03
L16370.06
5-Methylnicotinsäure ≥97%
5-Methylnicotinsäure
Purity: 97%
CAS No.: 3222-49-9
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Melting Point: 210-215°
Beilstein Reference: 112288
Formel:
C₇H₇NO₂ Molecular Weight: 137.14 g/mol Schmelzpunkt: 210…215 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00829036 CAS-Nummer: 3222-49-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...