5-Methylnicotinsäure
Lieferant: Thermo Scientific
Synonyme:
5-Methyl-3-pyridincarbonsäure, 5-Methylpyridin-3-carbonsäure
BTB09653.250
BTB09653.1
MAYBBTB09653.250EA
0
EUR
MAYBBTB09653.250
MAYBBTB09653.1
5-Methylnicotinsäure
5-Methylnicotinsäure
Formel:
C₇H₇NO₂ |
MDL:
MFCD00829036 CAS-Nummer: 3222-49-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...