6-Aminonicotinsäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Amino-3-pyridincarbonsäure, 6-Aminopyridin-3-carbonsäure

A11223.06 A11223.14
A11223.06EA 93.4 EUR
A11223.06 A11223.14
6-Aminonicotinsäure 98%
6-Aminonicotinsäure

Purity: 98% CAS No.: 3167-49-5 Molecular Formula: C6H6N2O2 Formula Weight: 138.13 Melting Point: ca 320° dec. Beilstein Reference: 115992 Merck Reference: 14,456 MDL No.: MFCD00006326

Formel: C₆H₆N₂O₂
Molecular Weight: 138,13 g/mol
Schmelzpunkt: ∼ 320 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006326
CAS-Nummer: 3167-49-5
EINECS: 221-630-4
Merck Index: 14,456

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR