6-Aminonicotinsäure 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Amino-3-pyridincarbonsäure, 6-Aminopyridin-3-carbonsäure
202540050
202540250
202540010
ACRO202540050EA
91
EUR
ACRO202540050
ACRO202540250
ACRO202540010
6-Aminonicotinsäure 98%
6-Aminonicotinsäure
Formel:
C₆H₆N₂O₂ Molecular Weight: 138,13 g/mol Schmelzpunkt: >300 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006326 CAS-Nummer: 3167-49-5 EINECS: 221-630-4 Merck Index: 13,00455 |
Spezifikation Testergebnisse
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...