ε-Hexalacton ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Hexansäure-ε-lacton, Hexan-6-olid, Capronsäure-ε-lacton, 6-Hydroxyhexansäurelacton, ε-Caprolacton
A10299.0C
A10299.0E
A10299.22
A10299.36
A10299.0CEA
561
EUR
A10299.0C
A10299.0E
A10299.22
A10299.36
ε-Hexalacton ≥99%
ε-Hexalacton
Purity: 99%
CAS No.: 502-44-3
Molecular Formula: C6H10O2
Formula Weight: 114.14
Flash Point: 109°(228°F)
Physical Form: Liquid
Density: 1.078
Refractive Index: 1.4630
Appearance: Colourless
Melting Point: -2°
Boiling Point: 235-236°
Beilstein Reference: 106919
MDL No.: MFCD00003267
Formel:
C₆H₁₀O₂ Molecular Weight: 114.14 g/mol Siedepunkt: 235…236 °C Schmelzpunkt: –2 °C Dichte: 1.078 Flammpunkt: 109 °C (228 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003267 CAS-Nummer: 502-44-3 EINECS: 207-938-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...