6-Methylnicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Methyl-3-pyridincarbonsäure, 6-Methylpyridin-3-carbonsäure

284750-1G 284750-5G
SIAL284750-1GEA 26.2 EUR
SIAL284750-1G SIAL284750-5G
6-Methylnicotinsäure, Sigma-Aldrich®
6-Methylnicotinsäure
Formel: C₇H₇NO₂
Molecular Weight: 137,14 g/mol
Schmelzpunkt: 212 °C
MDL: MFCD00006341
CAS-Nummer: 3222-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR