4,4'-Dithiodianilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(4-aminophenyl)disulfid

M-Clarity™ Qualitätsstufe = MQ100

369462-25G 369462-5G
SIAL369462-25GEA 441 EUR
SIAL369462-25G SIAL369462-5G
4,4'-Dithiodianilin, Sigma-Aldrich®
4,4'-Dithiodianilin
Formel: C₁₂H₁₂N₂S₂
Molecular Weight: 248,37 g/mol
MDL: MFCD00007882
CAS-Nummer: 722-27-0
EINECS: 211-961-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR