Bis(4-methoxyphenyl)disulfid ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,2-Bis(4-methoxyphenyl)disulphane
B25399.03
B25399.06
B25399.03EA
49.9
EUR
B25399.03
B25399.06
Bis(4-methoxyphenyl)disulfid ≥97%
Bis(4-methoxyphenyl)disulfid
Purity: 97%
CAS No.: 5335-87-5
Molecular Formula: C14H14O2S2
Formula Weight: 278.39
Melting Point: 37-41°
Boiling Point: 200°/3mm
MDL No.: MFCD00041358
Formel:
C₁₄H₁₄O₂S₂ Molecular Weight: 278,4 g/mol Siedepunkt: 200 °C (3 mmHg) Schmelzpunkt: 37…41 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041358 CAS-Nummer: 5335-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...