trans-Anethol ≥98% (durch GC)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(E)-1-Methoxy-4-(prop-1-en-1-yl)benzene
A13482.14
A13482.22
A13482.36
A13482.14EA
22.6
EUR
A13482.14
A13482.22
A13482.36
trans-Anethol ≥98% (durch GC)
trans-Anethol
Stabilized
Purity: 98+%
CAS No.: 4180-23-8
Molecular Formula: C10H12O
Formula Weight: 148.21
Storage Temperature: 2°C to 8°C
Flash Point: 96°(205°F)
Refractive Index: 1.5610
Melting Point: 20-23°
Boiling Point: 232-235°
Beilstein Reference: 774229
Merck Reference: 14,643
MDL No.: MFCD00009284
Formel:
C₁₀H₁₂O Molecular Weight: 148,2 g/mol Siedepunkt: 232…235 °C Schmelzpunkt: 20…23 °C Dichte: 0,991 Flammpunkt: 96 °C (205 °F) |
MDL:
MFCD00009284 CAS-Nummer: 4180-23-8 EINECS: 224-052-0 Merck Index: 14,643 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...