trans-Anethol 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(E)-1-Methoxy-4-(prop-1-en-1-yl)benzene
152531000
152530050
152535000
ACRO152531000EA
42.9
EUR
ACRO152531000
ACRO152530050
ACRO152535000
trans-Anethol 99%
trans-Anethol
Formel:
C₁₀H₁₂O Molecular Weight: 148,2 g/mol Siedepunkt: 236 °C (1013 hPa) Schmelzpunkt: 23 °C Dichte: 0,9875 g/cm³ Flammpunkt: 90 °C |
MDL:
MFCD00009284 CAS-Nummer: 4180-23-8 EINECS: 224-052-0 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid or solid |
Infrared spectrum | Conforms |
Refractive index | 1.5590 to 1.5630 (20°C, 589 nm) |
Density | (20/4°C) 0.986 to 0.989 |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...