Amino-functionalized silica gel, sphärisch 40-75 μm particle size, Supelco®
Lieferant: Merck
Achtung
Synonyme: Silica gel 110 NH2
79297-100G 79297-1KG
79297-100GEA 193 EUR
79297-100G 79297-1KG
Amino-functionalized silica gel, sphärisch 40-75 μm particle size, Supelco®
Amino-functionalized silica gel
Lagertemperatur: Raumtemperatur
CAS-Nummer: 126850-14-4