2-(2H-Benzo[d][1,2,3]triazol-2-yl)-6-(sec-butyl)-4-(tert-butyl)phenol

Lieferant: LGC Standards PROMOCHEM
Achtung

TRCB206645-25 TRCB206645-50
PROOTRCB206645-25EA 278 EUR
PROOTRCB206645-25 PROOTRCB206645-50
2-(2H-Benzo[d][1,2,3]triazol-2-yl)-6-(sec-butyl)-4-(tert-butyl)phenol
2-(2H-Benzo[d][1,2,3]triazol-2-yl)-6-(sec-butyl)-4-(tert-butyl)phenol
Formel: C₂₀H₂₅N₃O
Molecular Weight: 323,43 g/mol
MDL: MFCD09751272
CAS-Nummer: 36437-37-3

Bestellen

Spezifikation Testergebnisse

Appearance Pale Yellow to Light Yellow Solid
NMR Conforms
HPLC Purity 97.52% (310 nm)
MS Conforms
FT-IR Conforms
Specific Rotation −0.5° (c = 0.2, Chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR