D-Alanin

Lieferant: BACHEM BIOCHEMICA

Synonyme: (R)-(-)-2-Aminopropionic acid, Alanin, D-α-Alanin, D(-)-Alanin, (R)-(-)-Alanin, H-D-Ala-OH, α-Alanin, (R)-(-)-2-Aminopropansäure, D(-)-2-Aminopropionic acid, (R)-(-)-2-Aminopropionsäure

4029158.0025 4029158.0100 4029158.0250
BACE4029158.0025EA 0 EUR
BACE4029158.0025 BACE4029158.0100 BACE4029158.0250
D-Alanin
D-Alanin
Formel: C₃H₇NO₂
Molecular Weight: 89.09 g/mol
Siedepunkt: 189
Schmelzpunkt: 79
MDL: MFCD00008077
CAS-Nummer: 338-69-2
EINECS: 206-418-1
Merck Index: 13,00203

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR