D-α-Phenylalanin 99+%

Lieferant: Thermo Scientific

Synonyme: D-Phenylalanin, (R)-(+)-2-Amino-3-phenylpropansäure, 3-Phenyl-D-alanin, H-D-Phe-OH, 3-Phenyl-D(+)-alanin, (R)-(+)-2-Amino-3-phenylpropionsäure, (R)-(+)-2-Amino-3-phenylpropionic acid

172040250 172041000
ACRO172040250EA 145 EUR
ACRO172040250 ACRO172041000
D-α-Phenylalanin 99+%
D-α-Phenylalanin
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Siedepunkt: 295 °C (1013 hPa)
Schmelzpunkt: 273…276 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004270
CAS-Nummer: 673-06-3
EINECS: 211-603-5
Merck Index: 12,07425

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
Specific optical rotation +32° to +35° (20°C, 589 nm) (c=2, water)
Titration with HClO4 ≥99.0 %
Arsenic (As) ≤3 ppm
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR